1-Methyl-4-(2-pyrrolidinylmethyl)piperazine dihydrochloride

Supplier: Matrix Scientific

Synonyms: 1-Methyl-4-(pyrrolidin-2-ylmethyl)piperazine dihydrochloride

051706-500MG 051706-2.5G
101896-716EA 384.62 USD
101896-716 101896-718
1-Methyl-4-(2-pyrrolidinylmethyl)piperazine dihydrochloride
1-Methyl-4-(2-pyrrolidinylmethyl)piperazine dihydrochloride

Hazard: Irritant

Formula: C₁₀H₂₃Cl₂N₃
MW: 256.22 g/mol
MDL Number: MFCD13561758
CAS Number: 1219960-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR