3-Methyl-1-(2-pyrrolidinylmethyl)piperidine dihydrochloride

Supplier: Matrix Scientific

051704-500MG 051704-2.5G
101896-708EA 364.62 USD
101896-708 101896-710
3-Methyl-1-(2-pyrrolidinylmethyl)piperidine dihydrochloride
3-Methyl-1-(2-pyrrolidinylmethyl)piperidine dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₄Cl₂N₂
MDL Number: MFCD13561756
CAS Number: 1220027-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR