2-Methyl-1-(2-pyrrolidinylmethyl)piperidine dihydrochloride

Supplier: Matrix Scientific

051703-500MG 051703-2.5G
101896-704EA 364.62 USD
101896-704 101896-706
2-Methyl-1-(2-pyrrolidinylmethyl)piperidine dihydrochloride
2-Methyl-1-(2-pyrrolidinylmethyl)piperidine dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₄Cl₂N₂
MW: 255.23 g/mol
MDL Number: MFCD13561755
CAS Number: 1220027-14-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR