N-Allyl-N-(2-pyrrolidinylmethyl)-2-propen-1-amine dihydrochloride
Supplier: Matrix Scientific
051702-500MG
051702-2.5G
101896-700EA
364.62
USD
101896-700
101896-702
N-Allyl-N-(2-pyrrolidinylmethyl)-2-propen-1-amine dihydrochloride
N-Allyl-N-(2-pyrrolidinylmethyl)-2-propen-1-amine dihydrochloride
Hazard: Irritant
Formula:
C₁₁H₂₂Cl₂N₂ |
MDL Number:
MFCD13561754 CAS Number: 1219964-65-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...