2-[Ethyl(2-pyrrolidinylmethyl)amino]-1-ethanol dihydrochloride

Supplier: Matrix Scientific

051701-500MG 051701-2.5G
101896-696EA 364.62 USD
101896-696 101896-698
2-[Ethyl(2-pyrrolidinylmethyl)amino]-1-ethanol dihydrochloride
2-[Ethyl(2-pyrrolidinylmethyl)amino]-1-ethanol dihydrochloride

Hazard: Irritant

Formula: C₉H₂₂Cl₂N₂O
MW: 245.19 g/mol
MDL Number: MFCD13561753
CAS Number: 1219964-04-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR