N-Methyl-N-(2-pyrrolidinylmethyl)-1-butanamine dihydrochloride
Supplier: Matrix Scientific
051700-500MG
051700-2.5G
101896-692EA
364.62
USD
101896-692
101896-694
N-Methyl-N-(2-pyrrolidinylmethyl)-1-butanamine dihydrochloride
N-Methyl-N-(2-pyrrolidinylmethyl)-1-butanamine dihydrochloride
Hazard: Irritant
Formula:
C₁₀H₂₄Cl₂N₂ |
MDL Number:
MFCD13561752 CAS Number: 1220027-32-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...