1-(2-Pyrrolidinylmethyl)-3-pyrrolidinol dihydrochloride
Supplier: Matrix Scientific
051699-500MG
051699-2.5G
101896-688EA
364.62
USD
101896-688
101896-690
1-(2-Pyrrolidinylmethyl)-3-pyrrolidinol dihydrochloride
1-(2-Pyrrolidinylmethyl)-3-pyrrolidinol dihydrochloride
Hazard: Irritant
Formula:
C₉H₂₀Cl₂N₂O MW: 243.18 g/mol |
MDL Number:
MFCD13561751 CAS Number: 1220019-30-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...