1-(2-Pyrrolidinylmethyl)piperidine dihydrochloride

Supplier: Matrix Scientific

051697-500MG 051697-2.5G
101896-680EA 364.62 USD
101896-680 101896-682
1-(2-Pyrrolidinylmethyl)piperidine dihydrochloride
1-(2-Pyrrolidinylmethyl)piperidine dihydrochloride

Hazard: Irritant

Formula: C₁₀H₂₂Cl₂N₂
MW: 241.21 g/mol
MDL Number: MFCD13561749
CAS Number: 1220027-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR