2-[Methyl(2-pyrrolidinylmethyl)amino]-1-ethanol dihydrochloride
Supplier: Matrix Scientific
051696-500MG
051696-2.5G
101896-676EA
166.15
USD
101896-676
101896-678
2-[Methyl(2-pyrrolidinylmethyl)amino]-1-ethanol dihydrochloride
2-[Methyl(2-pyrrolidinylmethyl)amino]-1-ethanol dihydrochloride
Hazard: Irritant
Formula:
C₈H₂₀Cl₂N₂O MW: 231.17 g/mol |
MDL Number:
MFCD13561748 CAS Number: 1220019-13-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...