N-Ethyl-N-(2-pyrrolidinylmethyl)-1-ethanamine dihydrochloride
Supplier: Matrix Scientific
051695-500MG
051695-2.5G
101896-672EA
364.62
USD
101896-672
101896-674
N-Ethyl-N-(2-pyrrolidinylmethyl)-1-ethanamine dihydrochloride
N-Ethyl-N-(2-pyrrolidinylmethyl)-1-ethanamine dihydrochloride
Hazard: Irritant
Formula:
C₉H₂₂Cl₂N₂ |
MDL Number:
MFCD13561747 CAS Number: 1220027-24-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...