1-(2-Pyrrolidinylmethyl)pyrrolidine dihydrochloride
Supplier: Matrix Scientific
051694-500MG
051694-2.5G
101896-668EA
364.62
USD
101896-668
101896-670
1-(2-Pyrrolidinylmethyl)pyrrolidine dihydrochloride
1-(2-Pyrrolidinylmethyl)pyrrolidine dihydrochloride
Hazard: Irritant
Formula:
C₉H₂₀Cl₂N₂ |
MDL Number:
MFCD13561746 CAS Number: 1219963-95-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...