N,N-Dimethyl(2-pyrrolidinyl)methanamine dihydrochloride

Supplier: Matrix Scientific

051693-500MG 051693-2.5G
101896-664EA 364.62 USD
101896-664 101896-666
N,N-Dimethyl(2-pyrrolidinyl)methanamine dihydrochloride
N,N-Dimethyl(2-pyrrolidinyl)methanamine dihydrochloride

Hazard: Irritant

Formula: C₇H₁₈Cl₂N₂
MDL Number: MFCD13561745
CAS Number: 224309-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR