N-Methyl(2-piperidinyl)-N-(tetrahydro-2H-pyran-4-ylmethyl)methanamine dihydrochloride
Supplier: Matrix Scientific
051692-500MG
051692-2.5G
101896-660EA
364.62
USD
101896-660
101896-662
N-Methyl(2-piperidinyl)-N-(tetrahydro-2H-pyran-4-ylmethyl)methanamine dihydrochloride
N-Methyl(2-piperidinyl)-N-(tetrahydro-2H-pyran-4-ylmethyl)methanamine dihydrochloride
Hazard: Irritant
Formula:
C₁₃H₂₈Cl₂N₂O MW: 299.29 g/mol |
MDL Number:
MFCD13561744 CAS Number: 1219963-99-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...