N-Benzyl-N-(2-piperidinylmethyl)-1-ethanamine dihydrochloride

Supplier: Matrix Scientific

051691-500MG 051691-2.5G
101896-656EA 364.62 USD
101896-656 101896-658
N-Benzyl-N-(2-piperidinylmethyl)-1-ethanamine dihydrochloride
N-Benzyl-N-(2-piperidinylmethyl)-1-ethanamine dihydrochloride

Hazard: Irritant

Formula: C₁₅H₂₆Cl₂N₂
MDL Number: MFCD13561743
CAS Number: 1220019-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR