1-(2-Piperidinylmethyl)indoline dihydrochloride

Supplier: Matrix Scientific

051689-500MG 051689-2.5G
101896-648EA 364.62 USD
101896-648 101896-650
1-(2-Piperidinylmethyl)indoline dihydrochloride
1-(2-Piperidinylmethyl)indoline dihydrochloride

Hazard: Irritant

Formula: C₁₄H₂₂Cl₂N₂
MDL Number: MFCD13561741
CAS Number: 1220021-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR