2-(2-Piperidinylmethyl)-1,2,3,4-tetrahydroisoquinoline dihydrochloride

Supplier: Matrix Scientific

051688-500MG 051688-2.5G
101896-644EA 364.62 USD
101896-644 101896-646
2-(2-Piperidinylmethyl)-1,2,3,4-tetrahydroisoquinoline dihydrochloride
2-(2-Piperidinylmethyl)-1,2,3,4-tetrahydroisoquinoline dihydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄Cl₂N₂
MW: 303.28 g/mol
MDL Number: MFCD13561740
CAS Number: 1219964-57-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR