1-(2-Piperidinylmethyl)azepane dihydrochloride

Supplier: Matrix Scientific

051679-500MG 051679-2.5G
101896-608EA 364.62 USD
101896-608 101896-610
1-(2-Piperidinylmethyl)azepane dihydrochloride
1-(2-Piperidinylmethyl)azepane dihydrochloride

Hazard: Irritant

Formula: C₁₂H₂₆Cl₂N₂
MW: 269.26 g/mol
MDL Number: MFCD13561731
CAS Number: 1219960-55-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR