2-{4-[2-(4-Piperidinyl)ethyl]-1-piperazinyl}-1-ethanol dihydrochloride

Supplier: Matrix Scientific

051630-500MG 051630-2.5G
101896-412EA 364.62 USD
101896-412 101896-414
2-{4-[2-(4-Piperidinyl)ethyl]-1-piperazinyl}-1-ethanol dihydrochloride
2-{4-[2-(4-Piperidinyl)ethyl]-1-piperazinyl}-1-ethanol dihydrochloride

Hazard: Irritant

Formula: C₁₃H₂₉Cl₂N₃O
MW: 314.3 g/mol
MDL Number: MFCD13561682
CAS Number: 1220033-14-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR