1-(3-Pyrrolidinyl)azepane dihydrochloride

Supplier: Matrix Scientific

051536-500MG 051536-2.5G
101896-036EA 364.62 USD
101896-036 101896-038
1-(3-Pyrrolidinyl)azepane dihydrochloride
1-(3-Pyrrolidinyl)azepane dihydrochloride

Hazard: Irritant

Formula: C₁₀H₂₂Cl₂N₂
MW: 241.21 g/mol
MDL Number: MFCD13561593
CAS Number: 1220020-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR