1-Benzyl-4-(3-pyrrolidinyl)piperazine dihydrochloride

Supplier: Matrix Scientific

051535-500MG 051535-2.5G
101896-032EA 364.62 USD
101896-032 101896-034
1-Benzyl-4-(3-pyrrolidinyl)piperazine dihydrochloride
1-Benzyl-4-(3-pyrrolidinyl)piperazine dihydrochloride

Hazard: Irritant

Formula: C₁₅H₂₅Cl₂N₃
MDL Number: MFCD13561592
CAS Number: 1220038-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR