4-Methyl-1-(3-pyrrolidinyl)piperidine dihydrochloride
Supplier: Matrix Scientific
051533-500MG
051533-2.5G
101896-024EA
364.62
USD
101896-024
101896-026
4-Methyl-1-(3-pyrrolidinyl)piperidine dihydrochloride
4-Methyl-1-(3-pyrrolidinyl)piperidine dihydrochloride
Hazard: Irritant
Formula:
C₁₀H₂₂Cl₂N₂ MW: 241.21 g/mol |
MDL Number:
MFCD13561590 CAS Number: 1219957-30-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...