3-Methyl-1-(3-pyrrolidinyl)piperidine dihydrochloride

Supplier: Matrix Scientific

051532-500MG 051532-2.5G
101896-020EA 364.62 USD
101896-020 101896-022
3-Methyl-1-(3-pyrrolidinyl)piperidine dihydrochloride
3-Methyl-1-(3-pyrrolidinyl)piperidine dihydrochloride

Hazard: Irritant

Formula: C₁₀H₂₂Cl₂N₂
MW: 241.21 g/mol
MDL Number: MFCD13561589
CAS Number: 1219957-56-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR