2-[Ethyl-(3-pyrrolidinyl)amino]-1-ethanol dihydrochloride

Supplier: Matrix Scientific

051529-500MG 051529-2.5G
101896-008EA 364.62 USD
101896-008 101896-010
2-[Ethyl-(3-pyrrolidinyl)amino]-1-ethanol dihydrochloride
2-[Ethyl-(3-pyrrolidinyl)amino]-1-ethanol dihydrochloride

Hazard: Irritant

Formula: C₈H₂₀Cl₂N₂O
MDL Number: MFCD13561586
CAS Number: 1219979-94-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR