1-(3-Pyrrolidinyl)-3-pyrrolidinol dihydrochloride

Supplier: Matrix Scientific

051527-500MG 051527-2.5G
101896-000EA 166.15 USD
101896-000 101896-002
1-(3-Pyrrolidinyl)-3-pyrrolidinol dihydrochloride
1-(3-Pyrrolidinyl)-3-pyrrolidinol dihydrochloride

Hazard: Irritant

Formula: C₈H₁₈Cl₂N₂O
MDL Number: MFCD13561584
CAS Number: 1220038-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR