4-(3-Pyrrolidinyl)morpholine dihydrochloride

Supplier: Matrix Scientific

051526-500MG 051526-2.5G
101895-996EA 364.62 USD
101895-996 101895-998
4-(3-Pyrrolidinyl)morpholine dihydrochloride
4-(3-Pyrrolidinyl)morpholine dihydrochloride

Hazard: Irritant

Formula: C₈H₁₈Cl₂N₂O
MW: 229.15 g/mol
MDL Number: MFCD13561583
CAS Number: 1219979-89-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR