1-(3-Pyrrolidinyl)piperidine dihydrochloride

Supplier: Matrix Scientific

051525-500MG 051525-2.5G
101895-992EA 364.62 USD
101895-992 101895-994
1-(3-Pyrrolidinyl)piperidine dihydrochloride
1-(3-Pyrrolidinyl)piperidine dihydrochloride

Hazard: Irritant

Formula: C₉H₂₀Cl₂N₂
MW: 227.18 g/mol
MDL Number: MFCD13561582
CAS Number: 184970-32-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR