2-[Methyl(3-pyrrolidinyl)amino]-1-ethanol dihydrochloride

Supplier: Matrix Scientific

051524-500MG 051524-2.5G
101895-988EA 364.62 USD
101895-988 101895-990
2-[Methyl(3-pyrrolidinyl)amino]-1-ethanol dihydrochloride
2-[Methyl(3-pyrrolidinyl)amino]-1-ethanol dihydrochloride

Hazard: Irritant

Formula: C₇H₁₈Cl₂N₂O
MW: 217.14 g/mol
MDL Number: MFCD13561581
CAS Number: 1219957-28-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR