N,N-Dimethylpyrrolidin-3-amine dihydrochloride

Supplier: Matrix Scientific

051521-500MG 051521-2.5G
101895-976EA 364.62 USD
101895-976 101895-978
N,N-Dimethylpyrrolidin-3-amine dihydrochloride
N,N-Dimethylpyrrolidin-3-amine dihydrochloride

Hazard: Irritant

Formula: C₆H₁₆Cl₂N₂
MW: 187.1106 g/mol
MDL Number: MFCD13561578
CAS Number: 50534-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR