N-Methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)-4-piperidinamine dihydrochloride
Supplier: Matrix Scientific
051520-500MG
051520-2.5G
101895-972EA
364.62
USD
101895-972
101895-974
N-Methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)-4-piperidinamine dihydrochloride
N-Methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)-4-piperidinamine dihydrochloride
Hazard: Irritant
Formula:
C₁₂H₂₆Cl₂N₂O |
MDL Number:
MFCD13561577 CAS Number: 1220020-18-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...