N-Benzyl-N-ethyl-4-piperidinamine dihydrochloride

Supplier: Matrix Scientific

051519-500MG 051519-2.5G
101895-968EA 364.62 USD
101895-968 101895-970
N-Benzyl-N-ethyl-4-piperidinamine dihydrochloride
N-Benzyl-N-ethyl-4-piperidinamine dihydrochloride

Hazard: Irritant

Formula: C₁₄H₂₄Cl₂N₂
MDL Number: MFCD05983655
CAS Number: 871112-87-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR