1-(4-Piperidinyl)-1,2,3,4-tetrahydroquinoline dihydrochloride

Supplier: Matrix Scientific

051518-500MG 051518-2.5G
101895-964EA 364.62 USD
101895-964 101895-966
1-(4-Piperidinyl)-1,2,3,4-tetrahydroquinoline dihydrochloride
1-(4-Piperidinyl)-1,2,3,4-tetrahydroquinoline dihydrochloride

Hazard: Irritant

Formula: C₁₄H₂₂Cl₂N₂
MDL Number: MFCD12197346
CAS Number: 1170419-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR