1-(4-Piperidinyl)indoline dihydrochloride
Supplier: Matrix Scientific
051517-500MG
051517-2.5G
101895-960EA
166.15
USD
101895-960
101895-962
1-(4-Piperidinyl)indoline dihydrochloride
1-(4-Piperidinyl)indoline dihydrochloride
Hazard: Irritant
Formula:
C₁₃H₂₀Cl₂N₂ MW: 275.22 g/mol |
MDL Number:
MFCD13561576 CAS Number: 1220020-04-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...