2-[4-(4-Piperidinyl)-1-piperazinyl]-1-ethanol dihydrochloride

Supplier: Matrix Scientific

051515-500MG 051515-2.5G
101895-952EA 166.15 USD
101895-952 101895-954
2-[4-(4-Piperidinyl)-1-piperazinyl]-1-ethanol dihydrochloride
2-[4-(4-Piperidinyl)-1-piperazinyl]-1-ethanol dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₅Cl₂N₃O
MDL Number: MFCD13186590
CAS Number: 1220035-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR