N-Cyclohexyl-N-ethyl-4-piperidinamine dihydrochloride

Supplier: Matrix Scientific

051514-500MG 051514-2.5G
101895-948EA 364.62 USD
101895-948 101895-950
N-Cyclohexyl-N-ethyl-4-piperidinamine dihydrochloride
N-Cyclohexyl-N-ethyl-4-piperidinamine dihydrochloride

Hazard: Irritant

Formula: C₁₃H₂₈Cl₂N₂
MDL Number: MFCD13561575
CAS Number: 1220038-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR