N-Benzyl-N-methyl-4-piperidinamine dihydrochloride

Supplier: Matrix Scientific

051513-500MG 051513-2.5G
101895-944EA 210.77 USD
101895-944 101895-946
N-Benzyl-N-methyl-4-piperidinamine dihydrochloride
N-Benzyl-N-methyl-4-piperidinamine dihydrochloride

Hazard: Irritant

Formula: C₁₃H₂₂Cl₂N₂
MDL Number: MFCD06799870
CAS Number: 871112-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR