4-(2-Ethyl-1-piperidinyl)piperidine dihydrochloride
Supplier: Matrix Scientific
051512-500MG
051512-2.5G
101895-940EA
364.62
USD
101895-940
101895-942
4-(2-Ethyl-1-piperidinyl)piperidine dihydrochloride
4-(2-Ethyl-1-piperidinyl)piperidine dihydrochloride
Hazard: Irritant
Formula:
C₁₂H₂₆Cl₂N₂ |
MDL Number:
MFCD13561574 CAS Number: 1220020-11-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...