N-Cyclohexyl-N-methyl-4-piperidinamine dihydrochloride
Supplier: Matrix Scientific
051511-500MG
051511-2.5G
101895-936EA
364.62
USD
101895-936
101895-938
N-Cyclohexyl-N-methyl-4-piperidinamine dihydrochloride
N-Cyclohexyl-N-methyl-4-piperidinamine dihydrochloride
Hazard: Irritant
Formula:
C₁₂H₂₆Cl₂N₂ |
MDL Number:
MFCD13561573 CAS Number: 548769-24-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...