N,N-Dipropyl-4-piperidinamine dihydrochloride

Supplier: Matrix Scientific

051510-500MG 051510-2.5G
101895-932EA 364.62 USD
101895-932 101895-934
N,N-Dipropyl-4-piperidinamine dihydrochloride
N,N-Dipropyl-4-piperidinamine dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₆Cl₂N₂
MDL Number: MFCD13561572
CAS Number: 1220037-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR