1-(4-Piperidinyl)-4-piperidinol dihydrochloride

Supplier: Matrix Scientific

051509-500MG 051509-2.5G
101895-928EA 364.62 USD
101895-928 101895-930
1-(4-Piperidinyl)-4-piperidinol dihydrochloride
1-(4-Piperidinyl)-4-piperidinol dihydrochloride

Hazard: Irritant

Formula: C₁₀H₂₂Cl₂N₂O
MW: 257.21 g/mol
MDL Number: MFCD07781142
CAS Number: 367501-46-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR