1-Benzyl-4-(4-piperidinyl)piperazine dihydrochloride
Supplier: Matrix Scientific
051507-500MG
051507-2.5G
101895-920EA
364.62
USD
101895-920
101895-922
1-Benzyl-4-(4-piperidinyl)piperazine dihydrochloride
1-Benzyl-4-(4-piperidinyl)piperazine dihydrochloride
Hazard: Irritant
Formula:
C₁₆H₂₇Cl₂N₃ |
MDL Number:
MFCD09027976 CAS Number: 868707-62-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...