1-Methyl-4-(4-piperidinyl)piperazine dihydrochloride

Supplier: Matrix Scientific

051506-500MG 051506-2.5G
101895-916EA 364.62 USD
101895-916 101895-918
1-Methyl-4-(4-piperidinyl)piperazine dihydrochloride
1-Methyl-4-(4-piperidinyl)piperazine dihydrochloride

Hazard: Irritant

Formula: C₁₀H₂₃Cl₂N₃
MDL Number: MFCD13561570
CAS Number: 1219979-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR