4-(4-Methyl-1-piperidinyl)piperidine dihydrochloride

Supplier: Matrix Scientific

051505-500MG 051505-2.5G
101895-912EA 166.15 USD
101895-912 101895-914
4-(4-Methyl-1-piperidinyl)piperidine dihydrochloride
4-(4-Methyl-1-piperidinyl)piperidine dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₄Cl₂N₂
MDL Number: MFCD13561569
CAS Number: 1211484-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR