4-(2-Methyl-1-piperidinyl)piperidine dihydrochloride

Supplier: Matrix Scientific

051503-500MG 051503-2.5G
101895-904EA 364.62 USD
101895-904 101895-906
4-(2-Methyl-1-piperidinyl)piperidine dihydrochloride
4-(2-Methyl-1-piperidinyl)piperidine dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₄Cl₂N₂
MW: 255.23 g/mol
MDL Number: MFCD12197146
CAS Number: 1181458-19-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR