N,N-Diallyl-4-piperidinamine dihydrochloride

Supplier: Matrix Scientific

051502-500MG 051502-2.5G
101895-900EA 364.62 USD
101895-900 101895-902
N,N-Diallyl-4-piperidinamine dihydrochloride
N,N-Diallyl-4-piperidinamine dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₂Cl₂N₂
MW: 253.22 g/mol
MDL Number: MFCD13561567
CAS Number: 1219979-80-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR