N-Butyl-N-methyl-4-piperidinamine dihydrochloride

Supplier: Matrix Scientific

051500-500MG 051500-2.5G
101895-892EA 364.62 USD
101895-892 101895-894
N-Butyl-N-methyl-4-piperidinamine dihydrochloride
N-Butyl-N-methyl-4-piperidinamine dihydrochloride

Hazard: Irritant

Formula: C₁₀H₂₄Cl₂N₂
MDL Number: MFCD13561565
CAS Number: 1220035-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR