1-(4-Piperidinyl)-3-pyrrolidinol dihydrochloride
Supplier: Matrix Scientific
Synonyms:
1-(Piperidin-4-yl)pyrrolidin-3-ol dihydrochloride
051499-500MG
051499-2.5G
101895-888EA
384.62
USD
101895-888
101895-890
1-(4-Piperidinyl)-3-pyrrolidinol dihydrochloride
1-(4-Piperidinyl)-3-pyrrolidinol dihydrochloride
Hazard: Irritant
Formula:
C₉H₂₀Cl₂N₂O MW: 243.18 g/mol |
MDL Number:
MFCD13561564 CAS Number: 1220019-95-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...