4-(1-Piperidinyl)piperidine dihydrochloride

Supplier: Matrix Scientific

051497-500MG 051497-2.5G
101895-880EA 364.62 USD
101895-880 101895-882
4-(1-Piperidinyl)piperidine dihydrochloride
4-(1-Piperidinyl)piperidine dihydrochloride

Hazard: Irritant

Formula: C₁₀H₂₂Cl₂N₂
MW: 241.21 g/mol
MDL Number: MFCD06658209
CAS Number: 4876-60-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR