N,N-Diethyl-4-piperidinamine dihydrochloride
Supplier: Matrix Scientific
051496-500MG
051496-2.5G
101895-876EA
33.85
USD
101895-876
101895-878
N,N-Diethyl-4-piperidinamine dihydrochloride
N,N-Diethyl-4-piperidinamine dihydrochloride
Hazard: Irritant
Formula:
C₉H₂₂Cl₂N₂ MW: 229.2 g/mol |
MDL Number:
MFCD13561563 CAS Number: 50534-25-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...