4-(1-Pyrrolidinyl)piperidine dihydrochloride

Supplier: Matrix Scientific

Synonyms: 4-(Pyrrolidin-1-yl)piperidine dihydrochloride

051495-500MG 051495-2.5G
101895-872EA 364.62 USD
101895-872 101895-874
4-(1-Pyrrolidinyl)piperidine dihydrochloride
4-(1-Pyrrolidinyl)piperidine dihydrochloride

Hazard: Irritant

Formula: C₉H₂₀Cl₂N₂
MW: 227.18 g/mol
Melting Pt: 335…338 °C
MDL Number: MFCD00035294
CAS Number: 4983-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR