4-(Dimethylamino)piperidine dihydrochloride ≥95%

Supplier: Matrix Scientific

051494-5G 051494-25G
101895-868EA 52.31 USD
101895-868 101895-870
4-(Dimethylamino)piperidine dihydrochloride ≥95%
4-(Dimethylamino)piperidine dihydrochloride

Hazard: Irritant

Formula: C₇H₁₈Cl₂N₂
MW: 201.1400 g/mol
MDL Number: MFCD00035296
CAS Number: 4876-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR